siRNA Details
| Gene Symbol: AU022252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012400.2 | NP_001012400.1 | 5 | 530 | ||
| GenBank mRNA | AK158349.1 | - | 5 | 488 | ||
| AK167445.1 | - | 5 | 492 | |||
| AK218497.1 | - | 2 | 75 | |||
| BC035522.1 | - | 5 | 513 | |||
| BC089614.1 | - | 5 | 512 | 6748514 | ||

AU022252
Genomic Map