siRNA Details
| Gene Symbol: BC049730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199150.1 | NP_954601.1 | 4 | 286 | ||
| GenBank mRNA | BC049730.1 | - | 4 | 286 | 6770790 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC049730| Gene Symbol: BC049730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199150.1 | NP_954601.1 | 4 | 286 | ||
| GenBank mRNA | BC049730.1 | - | 4 | 286 | 6770790 | |