siRNA Details
| Gene Symbol: Olfr533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011815.1 | NP_001011815.1 | 1 | 627 | ||
| GenBank mRNA | BC147452.1 | - | 1 | 672 | 9056094 | |
| BC147473.1 | - | 1 | 672 | 9056115 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr533| Gene Symbol: Olfr533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011815.1 | NP_001011815.1 | 1 | 627 | ||
| GenBank mRNA | BC147452.1 | - | 1 | 672 | 9056094 | |
| BC147473.1 | - | 1 | 672 | 9056115 | ||