siRNA Details
| Gene Symbol: Olfr426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206926.1 | NP_001193855.1 | 1 | 903 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr426| Gene Symbol: Olfr426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206926.1 | NP_001193855.1 | 1 | 903 | ||