siRNA Details
| Gene Symbol: Olfr517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011846.1 | NP_001011846.1 | 1 | 533 | ||
| GenBank mRNA | BC150830.1 | - | 1 | 556 | 9087741 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr517| Gene Symbol: Olfr517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011846.1 | NP_001011846.1 | 1 | 533 | ||
| GenBank mRNA | BC150830.1 | - | 1 | 556 | 9087741 | |