siRNA Details
| Gene Symbol: Olfr1505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011850.1 | NP_001011850.1 | 1 | 866 | ||
| GenBank mRNA | BC150832.1 | - | 1 | 946 | 9087743 | |
| Gene Symbol: Olfr1502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC106809.2 | - | 712 | |||
| BC106813.2 | - | 712 | ||||
| BC106814.2 | - | 712 | ||||

Olfr1505
Genomic Map