siRNA Details
| Gene Symbol: Olfr1505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011850.1 | NP_001011850.1 | 1 | 917 | ||
| GenBank mRNA | BC150832.1 | - | 1 | 997 | 9087743 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1505| Gene Symbol: Olfr1505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011850.1 | NP_001011850.1 | 1 | 917 | ||
| GenBank mRNA | BC150832.1 | - | 1 | 997 | 9087743 | |