siRNA Details
| Gene Symbol: Olfr1029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011852.2 | NP_001011852.2 | 1 | 743 | ||
| GenBank mRNA | BC150841.1 | - | 1 | 743 | 9087752 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1029| Gene Symbol: Olfr1029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011852.2 | NP_001011852.2 | 1 | 743 | ||
| GenBank mRNA | BC150841.1 | - | 1 | 743 | 9087752 | |