siRNA Details
| Gene Symbol: Olfr605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011854.2 | NP_001011854.2 | 1 | 427 | ||
| GenBank mRNA | BC147216.1 | - | 1 | 427 | 9056257 | |
| BC147217.1 | - | 1 | 427 | 9056245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr605| Gene Symbol: Olfr605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011854.2 | NP_001011854.2 | 1 | 427 | ||
| GenBank mRNA | BC147216.1 | - | 1 | 427 | 9056257 | |
| BC147217.1 | - | 1 | 427 | 9056245 | ||