siRNA Details
| Gene Symbol: Olfr180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011662.2 | NP_001011662.2 | 2 | 977 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr180| Gene Symbol: Olfr180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011662.2 | NP_001011662.2 | 2 | 977 | ||