siRNA Details
| Gene Symbol: Olfr1037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011532.2 | NP_001011532.1 | 1 | 383 | ||
| GenBank mRNA | BC147135.1 | - | 1 | 383 | 9056192 | |
| BC147136.1 | - | 1 | 383 | 9056181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1037| Gene Symbol: Olfr1037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011532.2 | NP_001011532.1 | 1 | 383 | ||
| GenBank mRNA | BC147135.1 | - | 1 | 383 | 9056192 | |
| BC147136.1 | - | 1 | 383 | 9056181 | ||