siRNA Details
| Gene Symbol: Otulin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013792.2 | NP_001013814.2 | 7 | 1469 | ||
| GenBank mRNA | AK150371.1 | - | 7 | 1472 | ||
| BC028541.1 | - | 2 | 669 | |||
| BC087945.1 | - | 4 | 1070 | 30505408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otulin| Gene Symbol: Otulin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013792.2 | NP_001013814.2 | 7 | 1469 | ||
| GenBank mRNA | AK150371.1 | - | 7 | 1472 | ||
| BC028541.1 | - | 2 | 669 | |||
| BC087945.1 | - | 4 | 1070 | 30505408 | ||