siRNA Details
| Gene Symbol: Sept11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009818.1 | NP_001009818.1 | 10 | 4950 | ||
| GenBank mRNA | AK031522.1 | - | 1 | 2284 | ||
| AK033173.1 | - | 1 | 2174 | |||
| AK075989.1 | - | 1 | 1741 | |||
| AK160319.1 | - | 1 | 1254 | |||
| BC031456.1 | - | 10 | 4857 | 4037515 | ||
| BC064466.1 | - | 10 | 4950 | 6825549 | ||

Sept11
Genomic Map