siRNA Details
Gene Symbol: Methig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024672.3 | NP_001019843.1 | 1, 2 | 513 | ||
GenBank mRNA | AB099516.1 | - | 1, 2 | 550 | ||
AK144032.1 | - | 1, 2 | 514 | |||
BC139180.1 | - | 1, 2 | 547 | 8862202 | ||
BC139182.1 | - | 1, 2 | 547 | 8862204 |
Gene Symbol: Higd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB110950.1 | - | 1, 2 | 564 |
