siRNA Details
| Gene Symbol: Tmem160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026938.1 | NP_081214.1 | 2, 3 | 484 | ||
| GenBank mRNA | AK007386.1 | - | 2, 3 | 334 | ||
| AK088338.1 | - | 2, 3 | 307 | |||
| AK166985.1 | - | 2, 3 | 188 | |||
| AK187896.1 | - | 1, 2 | 15 | |||
| BC028534.1 | - | 2, 3 | 484 | 1447954 | ||

Tmem160
Genomic Map