siRNA Details
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026940.4 | NP_081216.1 | 5 | 1103 | ||
GenBank mRNA | AK007503.1 | - | 5 | 1092 | ||
BC030425.1 | - | 3 | 1387 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026940.4 | NP_081216.1 | 5 | 1103 | ||
GenBank mRNA | AK007503.1 | - | 5 | 1092 | ||
BC030425.1 | - | 3 | 1387 |