siRNA Details
| Gene Symbol: C1qtnf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026979.5 | NP_081255.1 | 4 | 938 | ||
| GenBank mRNA | AK007683.1 | - | 4 | 939 | ||
| BC030324.1 | - | 4 | 939 | 5375090 | ||
| DQ002395.1 | - | 3 | 800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1qtnf2| Gene Symbol: C1qtnf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026979.5 | NP_081255.1 | 4 | 938 | ||
| GenBank mRNA | AK007683.1 | - | 4 | 939 | ||
| BC030324.1 | - | 4 | 939 | 5375090 | ||
| DQ002395.1 | - | 3 | 800 | |||