siRNA Details
| Gene Symbol: 1700007K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027037.2 | NP_081313.1 | 3 | 191 | ||
| GenBank mRNA | AK005730.1 | - | 3 | 186 | ||
| BC048596.1 | - | 3 | 191 | 6740370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700007K09Rik| Gene Symbol: 1700007K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027037.2 | NP_081313.1 | 3 | 191 | ||
| GenBank mRNA | AK005730.1 | - | 3 | 186 | ||
| BC048596.1 | - | 3 | 191 | 6740370 | ||