siRNA Details
| Gene Symbol: Ccdc115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027159.2 | NP_081435.1 | 5 | 1538 | ||
| GenBank mRNA | AK010002.1 | - | 5 | 1232 | ||
| AK151074.1 | - | 2 | 1745 | |||
| AK154179.1 | - | 5 | 1266 | |||
| AK162237.1 | - | 5 | 1151 | |||
| BC019430.1 | - | 5 | 1534 | 3491825 | ||

Ccdc115
Genomic Map