siRNA Details
| Gene Symbol: Prss32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027220.2 | NP_081496.2 | 4 | 665 | ||
| GenBank mRNA | AY266139.1 | - | 4 | 652 | ||
| BC024903.1 | - | 4 | 665 | 4208033 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss32| Gene Symbol: Prss32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027220.2 | NP_081496.2 | 4 | 665 | ||
| GenBank mRNA | AY266139.1 | - | 4 | 652 | ||
| BC024903.1 | - | 4 | 665 | 4208033 | ||