siRNA Details
| Gene Symbol: Polr2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027231.1 | NP_081507.1 | 5 | 419 | ||
| GenBank mRNA | AF176516.2 | - | 1 | 1853 | ||
| AK007913.1 | - | 5 | 434 | |||
| AK165600.1 | - | 5 | 446 | |||
| AK169434.1 | - | 5 | 435 | |||
| BC024419.1 | - | 5 | 419 | 4983359 | ||

Polr2f
Genomic Map