siRNA Details
Gene Symbol: Polr2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027231.1 | NP_081507.1 | 515 | |||
GenBank mRNA | BC024419.1 | - | 515 | 4983359 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Polr2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027231.1 | NP_081507.1 | 515 | |||
GenBank mRNA | BC024419.1 | - | 515 | 4983359 |