siRNA Details
Gene Symbol: 2010003K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027237.1 | NP_081513.1 | 2 | 994 | ||
GenBank mRNA | AK008072.1 | - | 2 | 972 | ||
BC021951.1 | - | 2 | 994 | 5102159 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2010003K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027237.1 | NP_081513.1 | 2 | 994 | ||
GenBank mRNA | AK008072.1 | - | 2 | 972 | ||
BC021951.1 | - | 2 | 994 | 5102159 |