siRNA Details
| Gene Symbol: 2010003K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027237.1 | NP_081513.1 | 1, 2 | 345 | ||
| GenBank mRNA | AK008072.1 | - | 1, 2 | 323 | ||
| BC021951.1 | - | 1, 2 | 345 | 5102159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010003K11Rik| Gene Symbol: 2010003K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027237.1 | NP_081513.1 | 1, 2 | 345 | ||
| GenBank mRNA | AK008072.1 | - | 1, 2 | 323 | ||
| BC021951.1 | - | 1, 2 | 345 | 5102159 | ||