siRNA Details
| Gene Symbol: Cyp2c66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011707.1 | NP_001011707.1 | 4, 5 | 681 | ||
| GenBank mRNA | AK008512.2 | - | 3, 4 | 507 | ||
| AY227734.1 | - | 4, 5 | 681 | |||
| BC147819.1 | - | 4, 5 | 678 | 9087536 | ||
| BC147824.1 | - | 4, 5 | 678 | 9087541 | ||

Cyp2c66
Genomic Map