siRNA Details
Gene Symbol: Zdhhc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027306.3 | NP_081582.1 | 1 | 820 | ||
GenBank mRNA | AK131706.1 | - | 1 | 794 | ||
BC049767.1 | - | 1 | 820 | 6774409 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zdhhc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027306.3 | NP_081582.1 | 1 | 820 | ||
GenBank mRNA | AK131706.1 | - | 1 | 794 | ||
BC049767.1 | - | 1 | 820 | 6774409 |