siRNA Details
| Gene Symbol: Fancd2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027633.3 | NP_081909.2 | 3 | 630 | ||
| GenBank mRNA | AK016461.1 | - | 3 | 571 | ||
| BC042706.1 | - | 1 | 286 | |||
| BC049660.1 | - | 3 | 630 | 6742399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fancd2os| Gene Symbol: Fancd2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027633.3 | NP_081909.2 | 3 | 630 | ||
| GenBank mRNA | AK016461.1 | - | 3 | 571 | ||
| BC042706.1 | - | 1 | 286 | |||
| BC049660.1 | - | 3 | 630 | 6742399 | ||