siRNA Details
| Gene Symbol: Amer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164705.1 | NP_001158177.1 | 2 | 1702 | ||
| NM_028113.3 | NP_082389.1 | 1 | 2080 | |||
| GenBank mRNA | AK011185.1 | - | 2 | 1637 | ||
| AK032651.1 | - | 1 | 2021 | |||
| AK139177.1 | - | 1 | 2027 | |||
| BC030356.1 | - | 1 | 795 | |||
| BC056350.1 | - | 2 | 1668 | 6415605 | ||

Amer2
Genomic Map