siRNA Details
| Gene Symbol: 1700001P01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC048607.1 | - | 3 | 519 | 6741651 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001P01Rik| Gene Symbol: 1700001P01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC048607.1 | - | 3 | 519 | 6741651 | |