siRNA Details
| Gene Symbol: Cyp2c65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028191.2 | NP_082467.2 | 2 | 244 | ||
| GenBank mRNA | AK008688.1 | - | 2 | 174 | ||
| AY227733.1 | - | 2 | 235 | |||
| BC051089.1 | - | 2 | 228 | |||
| BC117531.1 | - | 2 | 227 | 40106344 | ||
| BC118514.1 | - | 2 | 228 | 40106342 | ||

Cyp2c65
Genomic Map