siRNA Details
| Gene Symbol: Ccdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028804.1 | NP_083080.1 | 3 | 1728 | ||
| GenBank mRNA | AK009833.1 | - | 3 | 1572 | ||
| AK172247.1 | - | 2 | 1094 | |||
| BC058616.1 | - | 3 | 1728 | 5686139 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc3| Gene Symbol: Ccdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028804.1 | NP_083080.1 | 3 | 1728 | ||
| GenBank mRNA | AK009833.1 | - | 3 | 1572 | ||
| AK172247.1 | - | 2 | 1094 | |||
| BC058616.1 | - | 3 | 1728 | 5686139 | ||