siRNA Details
| Gene Symbol: Slc35f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029238.2 | NP_083514.1 | 8 | 1546 | ||
| GenBank mRNA | AB041538.1 | - | 9 | 1474 | ||
| AK034308.1 | - | 8 | 1548 | |||
| BC026501.1 | - | 1 | 419 | |||
| BC138981.1 | - | 8 | 1445 | 8862003 | ||
| BC145720.1 | - | 8 | 1445 | 40131023 | ||

Slc35f4
Genomic Map