siRNA Details
| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027164.1 | NP_081440.1 | 10 | 1719 | ||
| GenBank mRNA | AK009798.1 | - | 3 | 233 | ||
| AK165593.1 | - | 3 | 361 | |||
| BC050816.1 | - | 10 | 1719 | 6774506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc27| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027164.1 | NP_081440.1 | 10 | 1719 | ||
| GenBank mRNA | AK009798.1 | - | 3 | 233 | ||
| AK165593.1 | - | 3 | 361 | |||
| BC050816.1 | - | 10 | 1719 | 6774506 | ||