siRNA Details
| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027164.1 | NP_081440.1 | 11 | 2079 | ||
| GenBank mRNA | BC050816.1 | - | 11 | 2079 | 6774506 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc27| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027164.1 | NP_081440.1 | 11 | 2079 | ||
| GenBank mRNA | BC050816.1 | - | 11 | 2079 | 6774506 | |