siRNA Details
| Gene Symbol: PDSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020381.3 | NP_065114.3 | 5 | 1042 | ||
| XM_011535956.2 | XP_011534258.1 | 5 | 1140 | |||
| XM_011535957.2 | XP_011534259.1 | 5 | 1150 | |||
| XM_011535958.2 | XP_011534260.1 | 4 | 1006 | |||
| XM_011535959.2 | XP_011534261.1 | 5 | 1115 | |||
| XM_011535960.2 | XP_011534262.1 | 5 | 769 | |||
| XM_011535962.2 | XP_011534264.1 | 5 | 684 | |||
| XM_017011082.1 | XP_016866571.1 | 5 | 1062 | |||
| GenBank mRNA | AB210839.1 | - | 5 | 752 | ||
| AF254956.1 | - | 5 | 1013 | |||
| AK225629.1 | - | 5 | 1023 | |||
| AK301480.1 | - | 5 | 645 | |||
| AK315804.1 | - | 5 | 1023 | |||
| BC039906.1 | - | 5 | 1028 | 6066481 | ||

PDSS2
Genomic Map