siRNA Details
| Gene Symbol: Gm608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029889.2 | NP_001025060.1 | 8 | 6685 | ||
| GenBank mRNA | AK173333.1 | - | 1 | 3430 | ||
| BC141330.1 | - | 6 | 6474 | 9055942 | ||
| BC141331.1 | - | 6 | 6474 | 9055943 | ||

Gm608
Genomic Map