siRNA Details
| Gene Symbol: Otud7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025613.1 | NP_001020784.1 | 13 | 4205 | ||
| NM_001025614.1 | NP_001020785.1 | 12 | 4147 | |||
| GenBank mRNA | BC141397.1 | - | 12 | 4035 | 9056010 | |
| BC141398.1 | - | 12 | 4035 | 9056011 | ||

Otud7b
Genomic Map