siRNA Details
| Gene Symbol: Gmnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013761.2 | NP_001013783.1 | 6 | 1466 | ||
| NM_001285916.1 | NP_001272845.1 | 5 | 1560 | |||
| NM_001285918.1 | NP_001272847.1 | 5 | 1291 | |||
| GenBank mRNA | AK141059.1 | - | 5 | 1560 | ||
| AK160719.1 | - | 5 | 1260 | |||
| BC086669.1 | - | 5 | 1221 | 30639759 | ||

Gmnc
Genomic Map