siRNA Details
| Gene Symbol: Tmem171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025606.1 | NP_001020777.1 | 4 | 933 | ||
| GenBank mRNA | BC099490.1 | - | 4 | 933 | 30522938 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem171| Gene Symbol: Tmem171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025606.1 | NP_001020777.1 | 4 | 933 | ||
| GenBank mRNA | BC099490.1 | - | 4 | 933 | 30522938 | |