siRNA Details
Gene Symbol: Tmem171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025606.1 | NP_001020777.1 | 2 | 729 | ||
GenBank mRNA | BC099490.1 | - | 2 | 729 | 30522938 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025606.1 | NP_001020777.1 | 2 | 729 | ||
GenBank mRNA | BC099490.1 | - | 2 | 729 | 30522938 |