siRNA Details
| Gene Symbol: Unc13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029873.2 | NP_001025044.2 | 26 | 3303 | ||
| GenBank mRNA | AK147408.1 | - | 21 | 2816 | ||
| AY753536.1 | - | 26 | 3193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc13a| Gene Symbol: Unc13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029873.2 | NP_001025044.2 | 26 | 3303 | ||
| GenBank mRNA | AK147408.1 | - | 21 | 2816 | ||
| AY753536.1 | - | 26 | 3193 | |||