siRNA Details
| Gene Symbol: Urb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029876.1 | NP_001025047.1 | 3 | 3243 | ||
| GenBank mRNA | AK122209.1 | - | 1 | 2880 | ||
| AK133906.1 | - | 4 | 3557 | |||
| BC096020.1 | - | 1 | 1627 | |||
| BC151125.1 | - | 3 | 3322 | 9088038 | ||

Urb2
Genomic Map