siRNA Details
| Gene Symbol: Zfp942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199048.1 | NP_001185977.1 | 6 | 2278 | ||
| GenBank mRNA | AK136009.1 | - | 1 | 339 | ||
| BC096765.1 | - | 6 | 2264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp942| Gene Symbol: Zfp942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199048.1 | NP_001185977.1 | 6 | 2278 | ||
| GenBank mRNA | AK136009.1 | - | 1 | 339 | ||
| BC096765.1 | - | 6 | 2264 | |||