siRNA Details
| Gene Symbol: Odam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027128.2 | NP_081404.2 | 6, 7 | 538 | ||
| GenBank mRNA | AK009298.1 | - | 3, 4 | 242 | ||
| BC132394.1 | - | 3, 4 | 182 | 40130671 | ||
| BC132396.1 | - | 3, 4 | 181 | 40130673 | ||
| EF113909.1 | - | 6, 7 | 538 | |||

Odam
Genomic Map