siRNA Details
| Gene Symbol: Mex3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029890.2 | NP_001025061.2 | 2 | 2851 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mex3a| Gene Symbol: Mex3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029890.2 | NP_001025061.2 | 2 | 2851 | ||