siRNA Details
| Gene Symbol: Prr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163551.1 | NP_001157023.1 | 5 | 951 | ||
| NM_028648.3 | NP_082924.2 | 4 | 927 | |||
| GenBank mRNA | AK015291.1 | - | 4 | 928 | ||
| BC049645.1 | - | 5 | 961 | |||
| BC103787.1 | - | 5 | 962 | 30919261 | ||

Prr27
Genomic Map