siRNA Details
| Gene Symbol: Lonrf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028894.1 | NP_083170.1 | 11 | 2818 | ||
| GenBank mRNA | AK016522.1 | - | 11 | 2818 | ||
| AK167341.1 | - | 11 | 2877 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lonrf3| Gene Symbol: Lonrf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028894.1 | NP_083170.1 | 11 | 2818 | ||
| GenBank mRNA | AK016522.1 | - | 11 | 2818 | ||
| AK167341.1 | - | 11 | 2877 | |||