siRNA Details
Gene Symbol: Zfp943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025373.2 | NP_001020544.1 | 4 | 2066 | ||
GenBank mRNA | BC066031.1 | - | 4 | 1867 | 6418362 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025373.2 | NP_001020544.1 | 4 | 2066 | ||
GenBank mRNA | BC066031.1 | - | 4 | 1867 | 6418362 |