siRNA Details
Gene Symbol: Tmsb15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030106.2 | NP_084382.1 | 2 | 182 | ||
GenBank mRNA | AK018998.1 | - | 2 | 184 | ||
AK028086.1 | - | 2 | 146 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmsb15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030106.2 | NP_084382.1 | 2 | 182 | ||
GenBank mRNA | AK018998.1 | - | 2 | 184 | ||
AK028086.1 | - | 2 | 146 |