siRNA Details
| Gene Symbol: Srbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030133.3 | NP_084409.3 | 18 | 2355 | ||
| GenBank mRNA | AK144490.1 | - | 2 | 204 | ||
| BC038140.1 | - | 2 | 299 | |||
| BC100359.1 | - | 11 | 1219 | |||
| BC147429.1 | - | 2 | 232 | 9056482 | ||
| BC147430.1 | - | 2 | 232 | 9056479 | ||
| BC172066.1 | - | 3 | 339 | 9054352 | ||

Srbd1
Genomic Map